Assay Design



Gene Transcripts

Gene Symbol
DDHD2
Entrez Gene ID
23259
Gene Name
DDHD domain containing 2
Gene Aliases
SAMWD1, SPG54, iPLA(1)gamma
Chromosome Location
Chr.8: 38231343 - 38273442 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164232.1 NP_001157704.1 13 - 14 1812 85
NM_015214.2 NP_056029.2 13 - 14 1989 85
XM_005273454.2 XP_005273511.1 13 - 14 2137 85
XM_005273455.3 XP_005273512.1 13 - 14 2137 85
XM_005273456.3 XP_005273513.1 12 - 13 2047 85
XM_011544456.1 XP_011542758.1 13 - 14 2137 85
XM_017013252.1 XP_016868741.1 13 - 14 1946 85
XM_017013253.1 XP_016868742.1 13 - 14 2137 85
XM_017013254.1 XP_016868743.1 12 - 13 1726 85
XM_017013255.1 XP_016868744.1 11 - 12 1735 85
XM_017013256.1 XP_016868745.1 12 - 13 1825 85
GenBank mRNA AB018268.1 - 10 - 11 1199 85
AK023218.1 - 9 - 10 1018 85
AK056525.1 - 13 - 14 1891 85
AK094951.1 - 2 - 3 2402 85
AK125904.1 - 13 - 14 1812 85
AK127040.1 - 4 - 5 1340 85
AK299875.1 - 11 - 12 1396 85
BC010504.1 - 8 - 9 960 85

View Full Product Details