Assay Design



Gene Transcripts

Gene Symbol
DDHD2
Entrez Gene ID
23259
Gene Name
DDHD domain containing 2
Gene Aliases
SAMWD1, SPG54, iPLA(1)gamma
Chromosome Location
Chr.8: 38231343 - 38273442 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164232.1 NP_001157704.1 16 - 17 2248 106
NM_015214.2 NP_056029.2 16 - 17 2425 106
XM_005273454.2 XP_005273511.1 16 - 17 2573 106
XM_005273455.3 XP_005273512.1 16 - 17 2573 106
XM_005273456.3 XP_005273513.1 15 - 16 2483 106
XM_011544456.1 XP_011542758.1 16 - 17 2573 106
XM_017013252.1 XP_016868741.1 16 - 17 2382 106
XM_017013253.1 XP_016868742.1 16 - 17 2573 106
XM_017013254.1 XP_016868743.1 15 - 16 2162 106
XM_017013255.1 XP_016868744.1 14 - 15 2171 106
XM_017013256.1 XP_016868745.1 15 - 16 2261 106
GenBank mRNA AB018268.1 - 13 - 14 1635 106
AK023218.1 - 12 - 13 1454 106
AK056525.1 - 16 - 17 2327 106
AK094951.1 - 5 - 6 2838 106
AK125904.1 - 16 - 17 2248 106
AK127040.1 - 7 - 8 1776 106
BC010504.1 - 11 - 12 1396 106

View Full Product Details