Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000756.3 | NP_000747.1 | 1 - 2 | 353 | 131 |
XM_017013090.1 | XP_016868579.1 | 5 - 6 | 691 | 131 | |
XM_017013091.1 | XP_016868580.1 | 5 - 6 | 754 | 131 | |
XM_017013092.1 | XP_016868581.1 | 5 - 6 | 692 | 131 | |
XM_017013093.1 | XP_016868582.1 | 5 - 6 | 831 | 131 | |
GenBank mRNA | AK075431.1 | - | 1 - 2 | 299 | 131 |
BC002599.2 | - | 1 - 2 | 164 | 131 | |
BC011031.1 | - | 1 - 2 | 174 | 131 | |
CB992265.1 | - | 1 - 2 | 380 | 131 |