Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002596.3 | NP_002587.2 | 11 - 12 | 1419 | 56 |
NM_212502.2 | NP_997667.1 | 11 - 12 | 1337 | 56 | |
NM_212503.2 | NP_997668.1 | 11 - 12 | 1427 | 56 | |
XM_011509602.2 | XP_011507904.1 | 11 - 12 | 1477 | 56 | |
XM_017001423.1 | XP_016856912.1 | 11 - 12 | 1572 | 56 | |
GenBank mRNA | AB209307.1 | - | 9 - 10 | 1999 | 56 |
AK126879.1 | - | 8 - 9 | 1423 | 56 | |
AK293756.1 | - | 10 - 11 | 1701 | 56 | |
AK295272.1 | - | 9 - 10 | 867 | 56 | |
AK296320.1 | - | 11 - 12 | 1385 | 56 | |
AK308913.1 | - | 9 - 10 | 908 | 56 | |
AL161977.1 | - | 3 - 4 | 634 | 56 | |
AY353237.1 | - | 11 - 12 | 1252 | 56 | |
AY353238.1 | - | 11 - 12 | 1316 | 56 | |
BC000281.2 | - | 5 - 6 | 504 | 56 | |
BC011526.1 | - | 11 - 12 | 1272 | 56 | |
BC040529.1 | - | 3 - 4 | 730 | 56 | |
BT007299.1 | - | 10 - 11 | 1069 | 56 | |
X66362.1 | - | 8 - 9 | 793 | 56 |