Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002596.3 | NP_002587.2 | 15 - 16 | 1739 | 105 |
NM_212502.2 | NP_997667.1 | 15 - 16 | 1657 | 105 | |
NM_212503.2 | NP_997668.1 | 15 - 16 | 1747 | 105 | |
XM_011509602.2 | XP_011507904.1 | 15 - 16 | 1797 | 105 | |
XM_017001423.1 | XP_016856912.1 | 15 - 16 | 1892 | 105 | |
GenBank mRNA | AB209307.1 | - | 13 - 14 | 2319 | 105 |
AK126879.1 | - | 10 - 11 | 2530 | 105 | |
AK293756.1 | - | 14 - 15 | 2021 | 105 | |
AK295272.1 | - | 13 - 14 | 1187 | 105 | |
AK296320.1 | - | 15 - 16 | 1705 | 105 | |
AL161977.1 | - | 6 - 7 | 1649 | 105 | |
AY353237.1 | - | 15 - 16 | 1572 | 105 | |
AY353238.1 | - | 15 - 16 | 1636 | 105 | |
BC000281.2 | - | 9 - 10 | 824 | 105 | |
BC011526.1 | - | 15 - 16 | 1592 | 105 | |
BC040529.1 | - | 6 - 7 | 1745 | 105 | |
BT007299.1 | - | 14 - 15 | 1389 | 105 | |
X66362.1 | - | 12 - 13 | 1113 | 105 |