Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144758.2 | NP_001138230.1 | 9 - 10 | 2511 | 114 |
NM_001144759.2 | NP_001138231.1 | 9 - 10 | 2511 | 114 | |
NM_015157.3 | NP_055972.1 | 10 - 11 | 2734 | 114 | |
XM_005271468.3 | XP_005271525.1 | 9 - 10 | 3280 | 114 | |
XM_005271469.3 | XP_005271526.1 | 9 - 10 | 3280 | 114 | |
XM_005271470.3 | XP_005271527.1 | 9 - 10 | 3280 | 114 | |
XM_005271471.3 | XP_005271528.1 | 9 - 10 | 3280 | 114 | |
XM_005271472.3 | XP_005271529.1 | 9 - 10 | 3280 | 114 | |
XM_005271473.3 | XP_005271530.1 | 9 - 10 | 3280 | 114 | |
XM_005271474.4 | XP_005271531.1 | 9 - 10 | 3280 | 114 | |
XM_011542707.2 | XP_011541009.1 | 9 - 10 | 3280 | 114 | |
XM_011542715.2 | XP_011541017.1 | 4 - 5 | 1787 | 114 | |
XM_017017407.1 | XP_016872896.1 | 9 - 10 | 3277 | 114 | |
XM_017017410.1 | XP_016872899.1 | 9 - 10 | 3280 | 114 | |
GenBank mRNA | AB014538.2 | - | 9 - 10 | 2488 | 114 |
AB094090.1 | - | 10 - 11 | 2734 | 114 | |
AB177859.1 | - | 4 - 5 | 1615 | 114 | |
AK074070.2 | - | 9 - 10 | 2460 | 114 | |
AL832373.1 | - | 9 - 10 | 2468 | 114 | |
BC013031.2 | - | 3 - 4 | 497 | 114 | |
BC098586.1 | - | 9 - 10 | 2488 | 114 | |
BX648176.1 | - | 4 - 5 | 663 | 114 | |
CR749783.1 | - | 10 - 11 | 2647 | 114 |