Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144758.2 | NP_001138230.1 | 15 - 16 | 3248 | 58 |
NM_001144759.2 | NP_001138231.1 | 14 - 15 | 3107 | 58 | |
NM_015157.3 | NP_055972.1 | 16 - 17 | 3471 | 58 | |
XM_005271470.3 | XP_005271527.1 | 15 - 16 | 4017 | 58 | |
XM_005271471.3 | XP_005271528.1 | 15 - 16 | 4017 | 58 | |
XM_005271473.3 | XP_005271530.1 | 14 - 15 | 3876 | 58 | |
XM_005271474.4 | XP_005271531.1 | 14 - 15 | 3876 | 58 | |
XM_006718799.3 | XP_006718862.1 | 16 - 17 | 4146 | 58 | |
XM_006718800.3 | XP_006718863.1 | 16 - 17 | 4146 | 58 | |
XM_006718802.3 | XP_006718865.1 | 15 - 16 | 4005 | 58 | |
XM_006718803.3 | XP_006718866.1 | 15 - 16 | 4005 | 58 | |
XM_011542705.2 | XP_011541007.1 | 18 - 19 | 4461 | 58 | |
XM_011542706.2 | XP_011541008.1 | 18 - 19 | 4461 | 58 | |
XM_011542715.2 | XP_011541017.1 | 9 - 10 | 2383 | 58 | |
XM_017017408.1 | XP_016872897.1 | 17 - 18 | 4317 | 58 | |
XM_017017409.1 | XP_016872898.1 | 17 - 18 | 4290 | 58 | |
GenBank mRNA | AB014538.2 | - | 15 - 16 | 3225 | 58 |
AB094090.1 | - | 16 - 17 | 3471 | 58 | |
AK027510.1 | - | 6 - 7 | 736 | 58 | |
AK074070.2 | - | 14 - 15 | 3056 | 58 | |
AK074918.1 | - | 8 - 9 | 943 | 58 | |
AK125783.1 | - | 17 - 18 | 2475 | 58 | |
AK296345.1 | - | 4 - 5 | 616 | 58 | |
BC013031.2 | - | 8 - 9 | 1093 | 58 | |
BX648176.1 | - | 9 - 10 | 1259 | 58 |