Assay Design



Gene Transcripts

Gene Symbol
HAUS5
Entrez Gene ID
23354
Gene Name
HAUS augmin like complex subunit 5
Gene Aliases
KIAA0841, dgt5
Chromosome Location
Chr.19: 35612744 - 35625349 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015302.1 NP_056117.1 13 - 14 1234 86
XM_011526684.2 XP_011524986.1 12 - 13 1121 86
GenBank mRNA AB020648.1 - 13 - 14 1207 86
BC013947.2 - 13 - 14 1206 86
BC064390.1 - 12 - 13 1170 86
BC157882.1 - 13 - 14 1225 86
BC171905.1 - 13 - 14 1226 86

View Full Product Details