Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006947.1 | NP_001006948.1 | 2 - 3 | 435 | 80 |
NM_015054.1 | NP_055869.1 | 2 - 3 | 435 | 80 | |
XM_005268738.2 | XP_005268795.1 | 3 - 4 | 541 | 80 | |
XM_005268739.3 | XP_005268796.1 | 2 - 3 | 395 | 80 | |
XM_017019047.1 | XP_016874536.1 | 2 - 3 | 395 | 80 | |
XM_017019048.1 | XP_016874537.1 | 3 - 4 | 541 | 80 | |
XM_017019049.1 | XP_016874538.1 | 2 - 3 | 395 | 80 | |
XM_017019050.1 | XP_016874539.1 | 2 - 3 | 395 | 80 | |
XM_017019051.1 | XP_016874540.1 | 3 - 4 | 541 | 80 | |
GenBank mRNA | AK125813.1 | - | 2 - 3 | 395 | 80 |
BC014891.1 | - | 2 - 3 | 418 | 80 | |
BC127017.1 | - | 2 - 3 | 253 | 80 | |
CB988442.1 | - | 2 - 3 | 462 | 80 |