Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153267.4 | NP_694999.3 | 9 - 10 | 2024 | 78 |
NR_125850.1 | 9 - 10 | 2024 | 78 | ||
XM_011518471.1 | XP_011516773.1 | 9 - 10 | 2086 | 78 | |
XM_011518475.2 | XP_011516777.1 | 9 - 10 | 2086 | 78 | |
XM_017014587.1 | XP_016870076.1 | 9 - 10 | 2086 | 78 | |
GenBank mRNA | AK304040.1 | - | 9 - 10 | 1784 | 78 |
AK304531.1 | - | 9 - 10 | 1612 | 78 | |
BC015417.2 | - | 1 - 2 | 88 | 78 | |
BC016383.1 | - | 9 - 10 | 1742 | 78 | |
BC063634.1 | - | 9 - 10 | 2001 | 78 |