Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020801.2 | NP_065852.1 | 7 - 8 | 1423 | 140 |
GenBank mRNA | AB037797.1 | - | 7 - 8 | 1395 | 140 |
AK291753.1 | - | 7 - 8 | 1398 | 140 | |
BC015928.2 | - | 7 - 8 | 1390 | 140 | |
BC053619.1 | - | 7 - 8 | 1395 | 140 |