Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079675.2 | NP_001073143.1 | 8 - 9 | 1105 | 95 |
NM_001261437.1 | NP_001248366.1 | 7 - 8 | 994 | 95 | |
NM_001261438.1 | NP_001248367.1 | 7 - 8 | 823 | 95 | |
NM_001986.2 | NP_001977.1 | 8 - 9 | 934 | 95 | |
XM_017024349.1 | XP_016879838.1 | 8 - 9 | 896 | 95 | |
GenBank mRNA | AK290429.1 | - | 8 - 9 | 896 | 95 |
AK298872.1 | - | 6 - 7 | 740 | 95 | |
AK299019.1 | - | 7 - 8 | 785 | 95 | |
AK315961.1 | - | 7 - 8 | 885 | 95 | |
BC016623.1 | - | 8 - 9 | 876 | 95 | |
D12765.1 | - | 6 - 7 | 740 | 95 | |
U18018.1 | - | 8 - 9 | 1009 | 95 |