Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152278.3 | NP_689491.1 | 2 - 3 | 233 | 90 |
XM_017029655.1 | XP_016885144.1 | 2 - 3 | 174 | 90 | |
GenBank mRNA | AJ297363.1 | - | 2 - 3 | 189 | 90 |
BC005988.1 | - | 2 - 3 | 179 | 90 | |
BC016786.1 | - | 2 - 3 | 184 | 90 |