Assay Design



Gene Transcripts

Gene Symbol
DPH7
Entrez Gene ID
92715
Gene Name
diphthamide biosynthesis 7
Gene Aliases
C9orf112, RRT2, WDR85
Chromosome Location
Chr.9: 137554899 - 137578935 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138778.3 NP_620133.1 8 - 9 1108 85
XM_005266123.2 XP_005266180.1 9 - 10 1566 85
XM_005266126.3 XP_005266183.1 7 - 8 1339 85
XM_005266128.2 XP_005266185.1 9 - 10 1192 85
XM_005266129.2 XP_005266186.1 9 - 10 1195 85
XM_005266130.2 XP_005266187.1 7 - 8 963 85
XM_011519188.1 XP_011517490.1 7 - 8 1031 85
XM_011519189.1 XP_011517491.1 8 - 9 1209 85
XM_011519199.2 XP_011517501.1 8 - 9 1054 85
XM_011519200.1 XP_011517502.1 8 - 9 1057 85
XM_017015291.1 XP_016870780.1 9 - 10 1401 85
XM_017015292.1 XP_016870781.1 8 - 9 1471 85
XM_017015293.1 XP_016870782.1 8 - 9 1428 85
XM_017015294.1 XP_016870783.1 10 - 11 1381 85
XM_017015295.1 XP_016870784.1 8 - 9 1468 85
XM_017015296.1 XP_016870785.1 8 - 9 1307 85
XM_017015297.1 XP_016870786.1 10 - 11 1378 85
XM_017015298.1 XP_016870787.1 9 - 10 1661 85
XM_017015299.1 XP_016870788.1 9 - 10 918 85
XM_017015300.1 XP_016870789.1 9 - 10 1658 85
XM_017015301.1 XP_016870790.1 7 - 8 1331 85
XM_017015302.1 XP_016870791.1 7 - 8 2385 85
XM_017015303.1 XP_016870792.1 8 - 9 1566 85
XM_017015304.1 XP_016870793.1 7 - 8 1031 85
XM_017015305.1 XP_016870794.1 7 - 8 2362 85
XM_017015306.1 XP_016870795.1 8 - 9 1500 85
XM_017015307.1 XP_016870796.1 8 - 9 1497 85
XM_017015308.1 XP_016870797.1 6 - 7 2224 85
XM_017015309.1 XP_016870798.1 8 - 9 1224 85
GenBank mRNA AK075115.1 - 8 - 9 1109 85
BC003123.1 - 8 - 9 1047 85
BC017335.2 - 8 - 9 1067 85
BC040173.1 - 1 - 2 570 85

View Full Product Details