Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000947.4 | NP_000938.2 | 4 - 5 | 430 | 69 |
NM_001282487.1 | NP_001269416.1 | 4 - 5 | 430 | 69 | |
NM_001282488.1 | NP_001269417.1 | 4 - 5 | 461 | 69 | |
XM_017011004.1 | XP_016866493.1 | 4 - 5 | 430 | 69 | |
XM_017011005.1 | XP_016866494.1 | 4 - 5 | 430 | 69 | |
XM_017011006.1 | XP_016866495.1 | 4 - 5 | 430 | 69 | |
XM_017011007.1 | XP_016866496.1 | 3 - 4 | 345 | 69 | |
GenBank mRNA | AK223287.1 | - | 4 - 5 | 419 | 69 |
AK291915.1 | - | 4 - 5 | 417 | 69 | |
BC017833.1 | - | 4 - 5 | 430 | 69 | |
BC064931.1 | - | 4 - 5 | 415 | 69 | |
BI547222.1 | - | 4 - 5 | 466 | 69 | |
X74331.1 | - | 4 - 5 | 423 | 69 |