Assay Design



Gene Transcripts

Gene Symbol
LIN7B
Entrez Gene ID
64130
Gene Name
lin-7 homolog B, crumbs cell polarity complex component
Gene Aliases
LIN-7B, MALS-2, MALS2, VELI2
Chromosome Location
Chr.19: 49114339 - 49118460 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022165.2 NP_071448.1 3 - 4 276 92
GenBank mRNA AF311862.1 - 3 - 4 254 92
AY358744.1 - 3 - 4 276 92
BC027618.2 - 3 - 4 250 92

View Full Product Details