Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182924.3 | NP_891554.1 | 11 - 12 | 2333 | 104 |
XM_011515520.2 | XP_011513822.1 | 11 - 12 | 2357 | 104 | |
XM_011515522.2 | XP_011513824.1 | 11 - 12 | 2357 | 104 | |
XM_011515523.2 | XP_011513825.1 | 11 - 12 | 2363 | 104 | |
XM_011515524.2 | XP_011513826.1 | 5 - 6 | 557 | 104 | |
GenBank mRNA | AK027124.1 | - | 7 - 8 | 1697 | 104 |
AK074068.1 | - | 10 - 11 | 3734 | 104 | |
AK093249.1 | - | 2 - 3 | 1098 | 104 | |
AK126808.1 | - | 4 - 5 | 1315 | 104 | |
AL833704.2 | - | 10 - 11 | 2287 | 104 | |
BC037988.1 | - | 11 - 12 | 2320 | 104 |