Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031430.2 | NP_113618.2 | 3 - 4 | 728 | 98 |
XM_005256811.1 | XP_005256868.1 | 3 - 4 | 707 | 98 | |
XM_017025195.1 | XP_016880684.1 | 3 - 4 | 707 | 98 | |
GenBank mRNA | AF370391.1 | - | 3 - 4 | 670 | 97 |
AJ278711.1 | - | 3 - 4 | 728 | 98 | |
AJ404317.1 | - | 3 - 4 | 698 | 98 | |
AK055755.1 | - | 3 - 4 | 707 | 98 | |
AK314767.1 | - | 3 - 4 | 707 | 98 | |
BC004961.1 | - | 3 - 4 | 277 | 98 | |
BC031621.1 | - | 3 - 4 | 522 | 98 |