Assay Design
Gene Transcripts
- Gene Symbol
- ARHGEF7
- Entrez Gene ID
-
8874
- Gene Name
- Rho guanine nucleotide exchange factor 7
- Gene Aliases
-
BETA-PIX, COOL-1, COOL1, Nbla10314, P50, P50BP, P85, P85COOL1, P85SPR, PAK3, PIXB
- Chromosome Location
-
Chr.13: 111114619 - 111305734 on Build GRCh38
View Full Product Details