Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007055.3 | NP_008986.2 | 6 - 7 | 1022 | 109 |
GenBank mRNA | AF021351.1 | - | 7 - 8 | 889 | 109 |
AK300329.1 | - | 6 - 7 | 1002 | 109 | |
AY091459.1 | - | 6 - 7 | 889 | 109 | |
BC041089.1 | - | 6 - 7 | 983 | 109 |