Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004947.4 | NP_004938.1 | 33 - 34 | 3505 | 99 |
XM_005264914.3 | XP_005264971.1 | 33 - 34 | 3777 | 99 | |
XM_005264915.3 | XP_005264972.1 | 33 - 34 | 3777 | 99 | |
XM_005264917.3 | XP_005264974.1 | 33 - 34 | 3735 | 99 | |
XM_005264918.3 | XP_005264975.1 | 32 - 33 | 3407 | 99 | |
XM_006713008.3 | XP_006713071.1 | 33 - 34 | 3777 | 99 | |
XM_006713009.3 | XP_006713072.1 | 33 - 34 | 3777 | 99 | |
XM_006713010.3 | XP_006713073.1 | 33 - 34 | 3540 | 99 | |
XM_011533441.2 | XP_011531743.1 | 22 - 23 | 4386 | 99 | |
XM_011533443.2 | XP_011531745.1 | 9 - 10 | 2067 | 99 | |
XM_011533444.2 | XP_011531746.1 | 9 - 10 | 1991 | 99 | |
XM_011533445.2 | XP_011531747.1 | 9 - 10 | 2012 | 99 | |
XM_017005825.1 | XP_016861314.1 | 33 - 34 | 3556 | 99 | |
XM_017005826.1 | XP_016861315.1 | 33 - 34 | 3546 | 99 | |
XM_017005827.1 | XP_016861316.1 | 24 - 25 | 2697 | 99 | |
XM_017005828.1 | XP_016861317.1 | 8 - 9 | 1422 | 99 | |
GenBank mRNA | AB002297.1 | - | 28 - 29 | 3113 | 99 |
AY145303.1 | - | 33 - 34 | 3482 | 99 | |
AY254099.1 | - | 33 - 34 | 3505 | 99 |