Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004947.4 | NP_004938.1 | 35 - 36 | 3640 | 84 |
XM_005264914.3 | XP_005264971.1 | 35 - 36 | 3912 | 84 | |
XM_005264915.3 | XP_005264972.1 | 35 - 36 | 3912 | 84 | |
XM_005264916.3 | XP_005264973.1 | 34 - 35 | 3663 | 84 | |
XM_005264917.3 | XP_005264974.1 | 35 - 36 | 3870 | 84 | |
XM_005264918.3 | XP_005264975.1 | 34 - 35 | 3542 | 84 | |
XM_006713008.3 | XP_006713071.1 | 35 - 36 | 3912 | 84 | |
XM_006713009.3 | XP_006713072.1 | 35 - 36 | 3912 | 84 | |
XM_006713010.3 | XP_006713073.1 | 35 - 36 | 3675 | 84 | |
XM_011533441.2 | XP_011531743.1 | 24 - 25 | 4521 | 84 | |
XM_011533443.2 | XP_011531745.1 | 11 - 12 | 2202 | 84 | |
XM_011533444.2 | XP_011531746.1 | 11 - 12 | 2126 | 84 | |
XM_011533445.2 | XP_011531747.1 | 11 - 12 | 2147 | 84 | |
XM_017005825.1 | XP_016861314.1 | 35 - 36 | 3691 | 84 | |
XM_017005826.1 | XP_016861315.1 | 35 - 36 | 3681 | 84 | |
XM_017005827.1 | XP_016861316.1 | 26 - 27 | 2832 | 84 | |
XM_017005828.1 | XP_016861317.1 | 10 - 11 | 1557 | 84 | |
GenBank mRNA | AB002297.1 | - | 30 - 31 | 3248 | 84 |
AY145303.1 | - | 35 - 36 | 3617 | 84 | |
AY254099.1 | - | 35 - 36 | 3640 | 84 |