Assay Design



Gene Transcripts

Gene Symbol
MDN1
Entrez Gene ID
23195
Gene Name
midasin AAA ATPase 1
Gene Aliases
Rea1
Chromosome Location
Chr.6: 89642775 - 89828021 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014611.2 NP_055426.1 70 - 71 11913 87
XM_005248700.3 XP_005248757.1 31 - 32 5845 87
XM_006715405.3 XP_006715468.1 34 - 35 7391 87
XM_011535635.2 XP_011533937.1 70 - 71 11718 87
XM_011535636.2 XP_011533938.1 70 - 71 11883 87
GenBank mRNA AB002299.2 - 24 - 25 4569 87
AF503925.1 - 70 - 71 11842 87
AK125181.1 - 8 - 9 1713 87
CR627399.1 - 16 - 17 3368 87

View Full Product Details