Assay Design



Gene Transcripts

Gene Symbol
KDM6B
Entrez Gene ID
23135
Gene Name
lysine demethylase 6B
Gene Aliases
JMJD3
Chromosome Location
Chr.17: 7834210 - 7854800 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080424.1 NP_001073893.1 7 - 8 927 88
XM_005256549.3 XP_005256606.1 8 - 9 1079 88
XM_005256550.4 XP_005256607.1 7 - 8 921 88
XM_005256551.3 XP_005256608.1 7 - 8 960 88
XM_005256552.4 XP_005256609.1 7 - 8 1933 88
XM_006721483.3 XP_006721546.1 8 - 9 1024 88
XM_011523750.2 XP_011522052.1 8 - 9 2284 88
XM_011523752.2 XP_011522054.1 7 - 8 924 88
XM_017024381.1 XP_016879870.1 7 - 8 2370 88
GenBank mRNA AB002344.2 - 7 - 8 941 88

View Full Product Details