Assay Design



Gene Transcripts

Gene Symbol
ANKRD28
Entrez Gene ID
23243
Gene Name
ankyrin repeat domain 28
Gene Aliases
PITK, PPP1R65
Chromosome Location
Chr.3: 15667236 - 15859814 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195098.1 NP_001182027.1 9 - 10 1176 94
NM_001195099.1 NP_001182028.1 7 - 8 867 94
NM_015199.3 NP_056014.2 9 - 10 1132 94
XM_005264996.3 XP_005265053.1 9 - 10 2503 94
XM_011533540.2 XP_011531842.1 9 - 10 2512 94
XM_011533541.2 XP_011531843.1 9 - 10 2512 94
XM_011533542.2 XP_011531844.1 8 - 9 1316 94
XM_011533543.2 XP_011531845.1 8 - 9 2311 94
XM_011533545.2 XP_011531847.1 9 - 10 1839 94
XM_011533546.2 XP_011531848.1 9 - 10 1830 94
XM_011533547.2 XP_011531849.1 8 - 9 1788 94
XM_017006026.1 XP_016861515.1 10 - 11 1910 94
XM_017006027.1 XP_016861516.1 9 - 10 1859 94
XM_017006028.1 XP_016861517.1 9 - 10 2512 94
GenBank mRNA AB002377.2 - 9 - 10 1008 94
AK126888.1 - 9 - 10 1394 94
AK293770.1 - 9 - 10 1177 94
AK295813.1 - 8 - 9 1002 94
AY367056.1 - 7 - 8 867 94
BC106948.2 - 9 - 10 1206 94
BC113868.1 - 9 - 10 1195 94
BC114476.1 - 9 - 10 1206 94

View Full Product Details