Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015267.3 | NP_056082.2 | 19 - 20 | 3350 | 59 |
XM_011538061.2 | XP_011536363.1 | 19 - 20 | 4860 | 59 | |
XM_011538063.2 | XP_011536365.1 | 18 - 19 | 4725 | 59 | |
XM_011538069.2 | XP_011536371.1 | 19 - 20 | 3199 | 59 | |
XM_011538070.2 | XP_011536372.1 | 19 - 20 | 4109 | 59 | |
XM_017019080.1 | XP_016874569.1 | 20 - 21 | 4192 | 59 | |
XM_017019081.1 | XP_016874570.1 | 20 - 21 | 3255 | 59 | |
GenBank mRNA | AB006631.2 | - | 19 - 20 | 3349 | 59 |
BC151245.1 | - | 19 - 20 | 3360 | 59 |