Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009999.2 | NP_001009999.1 | 13 - 14 | 1698 | 106 |
NM_015013.3 | NP_055828.2 | 11 - 12 | 1626 | 106 | |
XM_005245786.2 | XP_005245843.1 | 12 - 13 | 1691 | 106 | |
XM_006710472.3 | XP_006710535.1 | 11 - 12 | 1631 | 106 | |
XM_006710473.2 | XP_006710536.1 | 12 - 13 | 1691 | 106 | |
XM_006710474.3 | XP_006710537.1 | 12 - 13 | 1691 | 106 | |
XM_017000716.1 | XP_016856205.1 | 12 - 13 | 1691 | 106 | |
XM_017000717.1 | XP_016856206.1 | 11 - 12 | 1631 | 106 | |
XM_017000718.1 | XP_016856207.1 | 10 - 11 | 1097 | 106 | |
XM_017000719.1 | XP_016856208.1 | 11 - 12 | 1241 | 106 | |
XM_017000720.1 | XP_016856209.1 | 10 - 11 | 1211 | 106 | |
GenBank mRNA | AB011173.1 | - | 11 - 12 | 1579 | 106 |
AK290328.1 | - | 11 - 12 | 1152 | 106 | |
AL833812.1 | - | 7 - 8 | 748 | 106 | |
BC016639.1 | - | 9 - 10 | 865 | 106 | |
BC025362.1 | - | 3 - 4 | 291 | 106 | |
BC040194.2 | - | 13 - 14 | 1653 | 106 | |
BC048134.1 | - | 11 - 12 | 1547 | 106 |