Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009999.2 | NP_001009999.1 | 20 - 21 | 2596 | 68 |
NM_015013.3 | NP_055828.2 | 18 - 19 | 2524 | 68 | |
XM_005245786.2 | XP_005245843.1 | 19 - 20 | 2589 | 68 | |
XM_006710472.3 | XP_006710535.1 | 18 - 19 | 2547 | 68 | |
XM_006710474.3 | XP_006710537.1 | 19 - 20 | 2607 | 68 | |
XM_017000718.1 | XP_016856207.1 | 17 - 18 | 2013 | 68 | |
XM_017000719.1 | XP_016856208.1 | 18 - 19 | 2139 | 68 | |
XM_017000720.1 | XP_016856209.1 | 17 - 18 | 2109 | 68 | |
GenBank mRNA | AB011173.1 | - | 18 - 19 | 2477 | 68 |
AK290328.1 | - | 18 - 19 | 2050 | 68 | |
AL833812.1 | - | 14 - 15 | 1646 | 68 | |
BC016639.1 | - | 16 - 17 | 1763 | 68 | |
BC025362.1 | - | 10 - 11 | 1189 | 68 | |
BC040194.2 | - | 20 - 21 | 2551 | 68 | |
BC048134.1 | - | 18 - 19 | 2445 | 68 | |
BQ006926.1 | - | 1 - 2 | 68 |