Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015295.2 | NP_056110.2 | 37 - 38 | 4908 | 81 |
XM_011525642.1 | XP_011523944.1 | 37 - 38 | 4908 | 81 | |
XM_011525643.2 | XP_011523945.1 | 37 - 38 | 4908 | 81 | |
XM_011525644.2 | XP_011523946.1 | 36 - 37 | 4816 | 81 | |
XM_017025684.1 | XP_016881173.1 | 38 - 39 | 5086 | 81 | |
GenBank mRNA | AB014550.1 | - | 11 - 12 | 1250 | 81 |
AK126324.1 | - | 22 - 23 | 2539 | 81 | |
AL080138.1 | - | 8 - 9 | 888 | 81 | |
CR627458.1 | - | 26 - 27 | 3132 | 81 |