Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021117.3 | NP_066940.2 | 1 - 2 | 297 | 63 |
GenBank mRNA | AB014558.1 | - | 1 - 2 | 201 | 63 |
AK291260.1 | - | 1 - 2 | 228 | 63 | |
AK302865.1 | - | 1 - 2 | 297 | 63 | |
BC041814.1 | - | 1 - 2 | 225 | 63 | |
EU219622.1 | - | 1 - 2 | 220 | 63 |