Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297595.1 | NP_001284524.1 | 6 - 7 | 881 | 58 |
NM_015260.3 | NP_056075.1 | 6 - 7 | 881 | 58 | |
GenBank mRNA | AB014600.2 | - | 6 - 7 | 868 | 58 |
AY706204.1 | - | 6 - 7 | 861 | 58 | |
BC025026.1 | - | 6 - 7 | 845 | 58 | |
BC063531.1 | - | 6 - 7 | 861 | 58 | |
BC110821.1 | - | 6 - 7 | 858 | 58 |