Assay Design



Gene Transcripts

Gene Symbol
DGKB
Entrez Gene ID
1607
Gene Name
diacylglycerol kinase beta
Gene Aliases
DAGK2, DGK, DGK-BETA
Chromosome Location
Chr.7: 14145049 - 14974777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004080.2 NP_004071.1 9 - 10 1019 85
NM_145695.2 NP_663733.1 9 - 10 1019 85
XM_005249628.2 XP_005249685.1 10 - 11 1547 85
XM_005249629.2 XP_005249686.1 10 - 11 1547 85
XM_005249630.2 XP_005249687.1 9 - 10 1060 85
XM_005249631.3 XP_005249688.1 9 - 10 1311 85
XM_011515153.2 XP_011513455.1 10 - 11 1385 85
XM_011515154.2 XP_011513456.1 10 - 11 1101 85
XM_011515156.2 XP_011513458.1 9 - 10 1060 85
XM_011515157.2 XP_011513459.1 10 - 11 1100 85
XM_011515158.2 XP_011513460.1 9 - 10 1236 85
XM_017011783.1 XP_016867272.1 10 - 11 1441 85
XM_017011784.1 XP_016867273.1 10 - 11 1101 85
XM_017011785.1 XP_016867274.1 9 - 10 1332 85
XM_017011786.1 XP_016867275.1 9 - 10 1315 85
XM_017011787.1 XP_016867276.1 9 - 10 1065 85
XM_017011788.1 XP_016867277.1 10 - 11 1384 85
XM_017011789.1 XP_016867278.1 10 - 11 1101 85
XM_017011790.1 XP_016867279.1 10 - 11 1384 85
XM_017011791.1 XP_016867280.1 10 - 11 1383 85
XM_017011792.1 XP_016867281.1 10 - 11 1383 85
GenBank mRNA AB018261.1 - 6 - 7 648 85
AK295668.1 - 10 - 11 1336 85
AK304719.1 - 9 - 10 1311 85
BC105005.1 - 9 - 10 899 85
BC143632.1 - 9 - 10 899 85
BC143633.1 - 9 - 10 899 85

View Full Product Details