Assay Design



Gene Transcripts

Gene Symbol
NFASC
Entrez Gene ID
23114
Gene Name
neurofascin
Gene Aliases
NF, NRCAML
Chromosome Location
Chr.1: 204828652 - 205022822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005388.2 NP_001005388.2 11 - 12 1467 65
NM_001005389.1 NP_001005389.2 11 - 12 1467 65
NM_001160331.1 NP_001153803.1 9 - 10 1262 65
NM_001160332.1 NP_001153804.1 11 - 12 1500 65
NM_001160333.1 NP_001153805.1 10 - 11 1449 65
NM_015090.3 NP_055905.2 11 - 12 1500 65
XM_005244989.3 XP_005245046.2 11 - 12 1580 65
XM_005244991.3 XP_005245048.2 11 - 12 1580 65
XM_005244992.4 XP_005245049.2 11 - 12 1580 65
XM_005244993.3 XP_005245050.2 10 - 11 1529 65
XM_005244997.3 XP_005245054.2 10 - 11 1529 65
XM_011509311.1 XP_011507613.1 10 - 11 1280 65
XM_011509312.1 XP_011507614.1 11 - 12 1580 65
XM_011509313.1 XP_011507615.1 10 - 11 1280 65
XM_011509314.1 XP_011507616.1 10 - 11 1529 65
XM_011509315.1 XP_011507617.1 9 - 10 1229 65
XM_011509316.1 XP_011507618.1 12 - 13 1477 65
XM_011509317.1 XP_011507619.1 10 - 11 1280 65
XM_011509318.2 XP_011507620.1 11 - 12 1502 65
XM_011509319.1 XP_011507621.1 11 - 12 1426 65
XM_011509320.2 XP_011507622.1 10 - 11 1451 65
XM_011509321.1 XP_011507623.1 10 - 11 1280 65
XM_011509322.1 XP_011507624.1 10 - 11 1280 65
XM_011509323.1 XP_011507625.1 10 - 11 1280 65
XM_011509324.1 XP_011507626.1 10 - 11 1280 65
XM_011509325.1 XP_011507627.1 10 - 11 1280 65
XM_011509326.1 XP_011507628.1 10 - 11 1280 65
XM_011509327.1 XP_011507629.1 10 - 11 1280 65
XM_011509328.1 XP_011507630.1 9 - 10 1229 65
XM_017000733.1 XP_016856222.1 11 - 12 1580 65
XM_017000734.1 XP_016856223.1 10 - 11 1529 65
XM_017000735.1 XP_016856224.1 10 - 11 1280 65
XM_017000736.1 XP_016856225.1 10 - 11 1280 65
XM_017000737.1 XP_016856226.1 10 - 11 1280 65
XM_017000738.1 XP_016856227.1 11 - 12 1502 65
XM_017000739.1 XP_016856228.1 11 - 12 1502 65
XM_017000740.1 XP_016856229.1 11 - 12 1502 65
XM_017000741.1 XP_016856230.1 8 - 9 1070 65
XM_017000742.1 XP_016856231.1 9 - 10 1216 65
XM_017000743.1 XP_016856232.1 10 - 11 1451 65
GenBank mRNA AB018299.3 - 10 - 11 1331 65
AB177861.1 - 4 - 5 565 65
AK090639.1 - 11 - 12 1426 65
AK097802.1 - 9 - 10 1277 65
AK295291.1 - 10 - 11 1428 65
AK299262.1 - 10 - 11 1428 65
AK308686.1 - 11 - 12 1580 65
BC008124.2 - 2 - 3 165 65
BC040674.1 - 12 - 13 1718 65
BC117674.2 - 10 - 11 1316 65
BC137013.1 - 9 - 10 1237 65
BC144454.1 - 9 - 10 1204 65
BC144455.1 - 10 - 11 1255 65

View Full Product Details