Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014957.2 | NP_055772.2 | 14 - 15 | 2574 | 113 |
XM_005250838.4 | XP_005250895.2 | 14 - 15 | 2465 | 113 | |
XM_005250839.2 | XP_005250896.2 | 14 - 15 | 2465 | 113 | |
XM_005250840.4 | XP_005250897.2 | 13 - 14 | 2309 | 113 | |
XM_005250841.3 | XP_005250898.2 | 13 - 14 | 2309 | 113 | |
XM_005250842.4 | XP_005250899.1 | 12 - 13 | 2326 | 113 | |
XM_011516933.1 | XP_011515235.1 | 14 - 15 | 2465 | 113 | |
XM_011516934.2 | XP_011515236.1 | 14 - 15 | 2465 | 113 | |
XM_011516937.1 | XP_011515239.1 | 14 - 15 | 2465 | 113 | |
XM_011516938.2 | XP_011515240.1 | 7 - 8 | 1620 | 113 | |
XM_011516939.2 | XP_011515241.1 | 5 - 6 | 1143 | 113 | |
XM_011516940.2 | XP_011515242.1 | 5 - 6 | 1120 | 113 | |
XM_017013241.1 | XP_016868730.1 | 14 - 15 | 2467 | 113 | |
XM_017013242.1 | XP_016868731.1 | 15 - 16 | 2828 | 113 | |
XM_017013243.1 | XP_016868732.1 | 6 - 7 | 1580 | 113 | |
GenBank mRNA | AB020677.2 | - | 9 - 10 | 1761 | 113 |
BC133023.1 | - | 12 - 13 | 2161 | 113 | |
BC144226.1 | - | 12 - 13 | 2161 | 113 |