Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020452.3 | NP_065185.1 | 24 - 25 | 2875 | 127 |
XM_005245355.2 | XP_005245412.1 | 23 - 24 | 3022 | 127 | |
XM_005245356.3 | XP_005245413.1 | 24 - 25 | 2975 | 127 | |
XM_017001871.1 | XP_016857360.1 | 23 - 24 | 3025 | 127 | |
XM_017001872.1 | XP_016857361.1 | 24 - 25 | 2974 | 127 | |
GenBank mRNA | AB032963.1 | - | 14 - 15 | 2007 | 127 |
AK304806.1 | - | 6 - 7 | 823 | 127 | |
AY302537.1 | - | 24 - 25 | 2875 | 127 | |
BC007837.2 | - | 10 - 11 | 1493 | 127 | |
BC041573.1 | - | 10 - 11 | 1493 | 127 |