Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001247996.1 | NP_001234925.1 | 24 - 25 | 2501 | 68 |
NM_018482.3 | NP_060952.2 | 23 - 24 | 2399 | 68 | |
XM_005250925.1 | XP_005250982.1 | 24 - 25 | 2408 | 68 | |
XM_006716563.3 | XP_006716626.1 | 24 - 25 | 2227 | 68 | |
XM_006716564.1 | XP_006716627.1 | 25 - 26 | 2386 | 68 | |
XM_006716565.3 | XP_006716628.1 | 23 - 24 | 2223 | 68 | |
XM_006716566.1 | XP_006716629.1 | 24 - 25 | 2408 | 68 | |
XM_011517052.2 | XP_011515354.1 | 24 - 25 | 2224 | 68 | |
XM_011517053.1 | XP_011515355.1 | 24 - 25 | 2430 | 68 | |
XM_017013467.1 | XP_016868956.1 | 22 - 23 | 2245 | 68 | |
XM_017013468.1 | XP_016868957.1 | 23 - 24 | 2469 | 68 | |
XM_017013469.1 | XP_016868958.1 | 23 - 24 | 2399 | 68 | |
GenBank mRNA | AB033075.1 | - | 17 - 18 | 1632 | 68 |
BC137135.1 | - | 22 - 23 | 2196 | 68 | |
BC144493.1 | - | 23 - 24 | 2298 | 68 | |
BX537768.1 | - | 16 - 17 | 1652 | 68 |