Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193465.1 | NP_001180394.1 | 3 - 4 | 1545 | 76 |
NM_001193466.1 | NP_001180395.1 | 2 - 3 | 1759 | 76 | |
NM_015443.3 | NP_056258.1 | 2 - 3 | 1461 | 76 | |
XM_006721823.1 | XP_006721886.1 | 3 - 4 | 1545 | 76 | |
XM_006721824.3 | XP_006721887.1 | 3 - 4 | 1729 | 76 | |
XM_011524628.2 | XP_011522930.1 | 3 - 4 | 1729 | 76 | |
XM_011524629.2 | XP_011522931.1 | 3 - 4 | 1729 | 76 | |
XM_011524630.2 | XP_011522932.1 | 3 - 4 | 1729 | 76 | |
XM_011524631.2 | XP_011522933.1 | 3 - 4 | 1730 | 76 | |
XM_017024488.1 | XP_016879977.1 | 3 - 4 | 1545 | 76 | |
XM_017024489.1 | XP_016879978.1 | 2 - 3 | 1656 | 76 | |
GenBank mRNA | AB033093.1 | - | 1 - 2 | 1382 | 76 |
AK291259.1 | - | 2 - 3 | 1522 | 76 | |
BC098376.1 | - | 2 - 3 | 1461 | 76 | |
BX538006.1 | - | 2 - 3 | 1762 | 76 |