Assay Design



Gene Transcripts

Gene Symbol
KANSL1
Entrez Gene ID
284058
Gene Name
KAT8 regulatory NSL complex subunit 1
Gene Aliases
CENP-36, KDVS, KIAA1267, MSL1v1, NSL1, hMSL1v1
Chromosome Location
Chr.17: 46029916 - 46225374 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193465.1 NP_001180394.1 6 - 7 1913 64
NM_001193466.1 NP_001180395.1 5 - 6 2127 64
NM_015443.3 NP_056258.1 5 - 6 1829 64
XM_006721823.1 XP_006721886.1 6 - 7 1913 64
XM_006721824.3 XP_006721887.1 6 - 7 2097 64
XM_011524628.2 XP_011522930.1 6 - 7 2097 64
XM_011524629.2 XP_011522931.1 5 - 6 1995 64
XM_011524630.2 XP_011522932.1 6 - 7 2097 64
XM_011524631.2 XP_011522933.1 6 - 7 2098 64
XM_011524632.2 XP_011522934.1 5 - 6 523 64
XM_017024488.1 XP_016879977.1 6 - 7 1913 64
XM_017024489.1 XP_016879978.1 4 - 5 1922 64
GenBank mRNA AB033093.1 - 4 - 5 1750 64
AK291259.1 - 5 - 6 1890 64
AL137317.1 - 1 - 2 71 64
BC098376.1 - 5 - 6 1829 64
BX538006.1 - 5 - 6 2130 64

View Full Product Details