Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112095312-112095847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDHD
Entrez Gene ID
6392
Gene Name
succinate dehydrogenase complex subunit D
Gene Aliases
CBT1, CII-4, CWS3, PGL, PGL1, QPs3, SDH4, cybS
UniGene
Hs.356270
Gene Chromosome Location
Chr.11: 112086824-112095801 on Build GRCh38
Transcripts
NM_001276503.1 NM_001276504.1 NM_001276506.1 NM_003002.3 NR_077060.1

SNP Detail

SNP ID Chromosome Location
rs763123482 Chr.11: 112095781-112095781 on Build GRCh38
rs889587088 Chr.11: 112095412-112095412 on Build GRCh38
rs201560447 Chr.11: 112095618-112095618 on Build GRCh38
rs992322574 Chr.11: 112095642-112095642 on Build GRCh38
rs764507748 Chr.11: 112095794-112095794 on Build GRCh38
rs886047701 Chr.11: 112095557-112095557 on Build GRCh38
rs199710271 Chr.11: 112095679-112095679 on Build GRCh38
rs945574833 Chr.11: 112095657-112095657 on Build GRCh38
rs200798663 Chr.11: 112095411-112095411 on Build GRCh38
rs540110996 Chr.11: 112095368-112095368 on Build GRCh38
rs201695244 Chr.11: 112095440-112095440 on Build GRCh38
rs555396316 Chr.11: 112095736-112095736 on Build GRCh38
rs201100300 Chr.11: 112095568-112095568 on Build GRCh38
rs548950317 Chr.11: 112095423-112095423 on Build GRCh38
rs201470807 Chr.11: 112095365-112095365 on Build GRCh38
rs952084175 Chr.11: 112095584-112095584 on Build GRCh38
rs142510105 Chr.11: 112095592-112095592 on Build GRCh38
rs201472512 Chr.11: 112095792-112095792 on Build GRCh38
rs17113461 Chr.11: 112095773-112095773 on Build GRCh38
rs11547884 Chr.11: 112095665-112095665 on Build GRCh38
rs200264243 Chr.11: 112095633-112095633 on Build GRCh38
rs959465427 Chr.11: 112095639-112095639 on Build GRCh38
rs981255032 Chr.11: 112095823-112095823 on Build GRCh38
rs1011512562 Chr.11: 112095337-112095337 on Build GRCh38
rs183686123 Chr.11: 112095747-112095747 on Build GRCh38
rs544286614 Chr.11: 112095769-112095769 on Build GRCh38
rs897002374 Chr.11: 112095785-112095785 on Build GRCh38
rs886047702 Chr.11: 112095708-112095708 on Build GRCh38
rs796854714 Chr.11: 112095668-112095668 on Build GRCh38
rs567931768 Chr.11: 112095712-112095712 on Build GRCh38
rs533163850 Chr.11: 112095361-112095361 on Build GRCh38
rs953640580 Chr.11: 112095374-112095374 on Build GRCh38
rs191837062 Chr.11: 112095732-112095732 on Build GRCh38
rs200088980 Chr.11: 112095444-112095444 on Build GRCh38
rs573776592 Chr.11: 112095753-112095753 on Build GRCh38
rs201388681 Chr.11: 112095531-112095531 on Build GRCh38
rs1007666445 Chr.11: 112095714-112095715 on Build GRCh38
rs889313831 Chr.11: 112095702-112095702 on Build GRCh38
rs915707804 Chr.11: 112095503-112095503 on Build GRCh38
rs951889323 Chr.11: 112095766-112095770 on Build GRCh38
rs11214079 Chr.11: 112095717-112095717 on Build GRCh38
rs965711232 Chr.11: 112095421-112095421 on Build GRCh38
rs919018696 Chr.11: 112095439-112095439 on Build GRCh38
rs693441 Chr.11: 112095583-112095583 on Build GRCh38
rs766039453 Chr.11: 112095473-112095473 on Build GRCh38
rs199841245 Chr.11: 112095555-112095555 on Build GRCh38
rs146261846 Chr.11: 112095502-112095502 on Build GRCh38
rs201939906 Chr.11: 112095631-112095631 on Build GRCh38
rs187116888 Chr.11: 112095462-112095462 on Build GRCh38
rs918120569 Chr.11: 112095644-112095644 on Build GRCh38
rs886047703 Chr.11: 112095785-112095785 on Build GRCh38
rs971737341 Chr.11: 112095422-112095423 on Build GRCh38
rs200753024 Chr.11: 112095339-112095339 on Build GRCh38
rs924916478 Chr.11: 112095701-112095701 on Build GRCh38
rs201421504 Chr.11: 112095635-112095635 on Build GRCh38
rs199954470 Chr.11: 112095532-112095532 on Build GRCh38
rs536971023 Chr.11: 112095734-112095734 on Build GRCh38
rs879511523 Chr.11: 112095484-112095484 on Build GRCh38
rs139418464 Chr.11: 112095549-112095549 on Build GRCh38
rs184654032 Chr.11: 112095398-112095398 on Build GRCh38
rs200893241 Chr.11: 112095545-112095545 on Build GRCh38

View Full Product Details