Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003683.2 | NP_001003683.1 | 2 - 3 | 294 | 57 |
NM_001258312.1 | NP_001245241.1 | 2 - 3 | 321 | 57 | |
NM_005019.4 | NP_005010.2 | 1 - 2 | 336 | 57 | |
XM_011511323.2 | XP_011509625.1 | 1 - 2 | 113 | 57 | |
XM_011511324.2 | XP_011509626.1 | 2 - 3 | 341 | 57 | |
XM_017004294.1 | XP_016859783.1 | 1 - 2 | 336 | 57 | |
XM_017004295.1 | XP_016859784.1 | 2 - 3 | 341 | 57 | |
XM_017004296.1 | XP_016859785.1 | 2 - 3 | 263 | 57 | |
XM_017004297.1 | XP_016859786.1 | 2 - 3 | 204 | 57 | |
XM_017004298.1 | XP_016859787.1 | 2 - 3 | 386 | 57 | |
XM_017004299.1 | XP_016859788.1 | 2 - 3 | 341 | 57 | |
XM_017004300.1 | XP_016859789.1 | 2 - 3 | 386 | 57 | |
XM_017004301.1 | XP_016859790.1 | 2 - 3 | 191 | 57 | |
GenBank mRNA | AF110236.1 | - | - | 234 | 57 |
AF110237.1 | - | - | 236 | 57 | |
AK294239.1 | - | 2 - 3 | 256 | 57 | |
BC022480.1 | - | 1 - 2 | 304 | 57 | |
DA128734.1 | - | 1 - 2 | 337 | 57 | |
U40370.1 | - | 2 - 3 | 187 | 57 |