Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282613.1 | NP_001269542.1 | 4 - 5 | 683 | 67 |
NM_001282614.1 | NP_001269543.1 | 6 - 7 | 945 | 67 | |
NM_032735.2 | NP_116124.2 | 6 - 7 | 945 | 67 | |
XM_006719256.3 | XP_006719319.1 | 5 - 6 | 733 | 67 | |
XM_011537961.1 | XP_011536263.1 | 4 - 5 | 553 | 67 | |
XM_011537962.2 | XP_011536264.1 | 3 - 4 | 316 | 67 | |
XM_011537963.1 | XP_011536265.1 | 3 - 4 | 324 | 67 | |
GenBank mRNA | AF440758.1 | - | 6 - 7 | 890 | 67 |
AK096459.1 | - | 6 - 7 | 946 | 67 | |
AY515706.1 | - | 5 - 6 | 718 | 67 | |
BC151138.1 | - | 4 - 5 | 552 | 67 |