Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017510.4 | NP_059980.2 | 2 - 3 | 285 | 92 |
GenBank mRNA | AF441399.2 | - | 2 - 3 | 237 | 92 |
AK298435.1 | - | 2 - 3 | 225 | 92 | |
AK313292.1 | - | 2 - 3 | 294 | 92 | |
BC001123.2 | - | 2 - 3 | 285 | 92 |