Assay Design



Gene Transcripts

Gene Symbol
DIP2A
Entrez Gene ID
23181
Gene Name
disco interacting protein 2 homolog A
Gene Aliases
C21orf106, DIP2
Chromosome Location
Chr.21: 46458942 - 46570900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146115.1 NP_001139587.1 17 - 18 2232 71
NM_001146116.1 NP_001139588.1 18 - 19 2349 71
NM_015151.3 NP_055966.2 18 - 19 2361 71
NM_206889.2 NP_996772.1 18 - 19 2361 71
NM_206890.2 NP_996773.1 18 - 19 2361 71
NM_206891.2 NP_996774.1 18 - 19 2361 71
XM_011529492.2 XP_011527794.1 19 - 20 2476 71
XM_011529501.1 XP_011527803.1 19 - 20 2476 71
XM_011529502.1 XP_011527804.1 19 - 20 2476 71
XM_017028291.1 XP_016883780.1 19 - 20 2476 71
XM_017028292.1 XP_016883781.1 19 - 20 2476 71
XM_017028293.1 XP_016883782.1 19 - 20 2473 71
XM_017028294.1 XP_016883783.1 18 - 19 2355 71
XM_017028295.1 XP_016883784.1 18 - 19 2352 71
XM_017028296.1 XP_016883785.1 18 - 19 2352 71
XM_017028297.1 XP_016883786.1 18 - 19 2344 71
XM_017028298.1 XP_016883787.1 17 - 18 2223 71
XM_017028299.1 XP_016883788.1 18 - 19 2221 71
XM_017028300.1 XP_016883789.1 19 - 20 2476 71
XM_017028301.1 XP_016883790.1 19 - 20 2476 71
XM_017028302.1 XP_016883791.1 17 - 18 2116 71
XM_017028303.1 XP_016883792.1 19 - 20 2476 71
XM_017028304.1 XP_016883793.1 19 - 20 2476 71
XM_017028305.1 XP_016883794.1 19 - 20 2476 71
GenBank mRNA AB273729.1 - 18 - 19 2199 71
AF432263.1 - 13 - 14 1571 71
AF490768.1 - 18 - 19 2254 71
AK056738.1 - 18 - 19 2253 71
AK091790.1 - 2 - 3 1399 71
AK303351.1 - 16 - 17 2176 71
BC033718.1 - 17 - 18 2215 71
BC038443.1 - 18 - 19 2360 71
CK821329.1 - 2 - 3 71

View Full Product Details