Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146116.1 | NP_001139588.1 | 32 - 33 | 4142 | 66 |
NM_015151.3 | NP_055966.2 | 32 - 33 | 4154 | 66 | |
XM_011529492.2 | XP_011527794.1 | 33 - 34 | 4269 | 66 | |
XM_017028291.1 | XP_016883780.1 | 33 - 34 | 4269 | 66 | |
XM_017028292.1 | XP_016883781.1 | 33 - 34 | 4269 | 66 | |
XM_017028293.1 | XP_016883782.1 | 33 - 34 | 4266 | 66 | |
XM_017028294.1 | XP_016883783.1 | 32 - 33 | 4148 | 66 | |
XM_017028295.1 | XP_016883784.1 | 32 - 33 | 4145 | 66 | |
XM_017028296.1 | XP_016883785.1 | 32 - 33 | 4145 | 66 | |
XM_017028297.1 | XP_016883786.1 | 32 - 33 | 4137 | 66 | |
XM_017028298.1 | XP_016883787.1 | 31 - 32 | 4016 | 66 | |
XM_017028299.1 | XP_016883788.1 | 32 - 33 | 4014 | 66 | |
XM_017028300.1 | XP_016883789.1 | 33 - 34 | 4269 | 66 | |
XM_017028301.1 | XP_016883790.1 | 33 - 34 | 4269 | 66 | |
XM_017028302.1 | XP_016883791.1 | 31 - 32 | 3909 | 66 | |
XM_017028303.1 | XP_016883792.1 | 33 - 34 | 4269 | 66 | |
XM_017028304.1 | XP_016883793.1 | 33 - 34 | 4269 | 66 | |
GenBank mRNA | AB273729.1 | - | 32 - 33 | 3992 | 66 |
AF490768.1 | - | 32 - 33 | 4047 | 66 |