Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015131.2 | NP_055946.1 | 2 - 3 | 416 | 114 |
GenBank mRNA | AK000836.1 | - | 1 - 2 | 112 | 114 |
BC033206.1 | - | 1 - 2 | 121 | 114 | |
D26488.1 | - | 2 - 3 | 389 | 114 | |
DA654485.1 | - | 2 - 3 | 416 | 114 | |
DA943836.1 | - | 2 - 3 | 392 | 114 |