Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146335.2 | NP_001139807.1 | 8 - 9 | 1507 | 77 |
NM_182767.5 | NP_877499.1 | 9 - 10 | 1984 | 77 | |
GenBank mRNA | AF265577.1 | - | 8 - 9 | 1489 | 77 |
AK001178.1 | - | 4 - 5 | 669 | 77 | |
AK291207.1 | - | 9 - 10 | 1916 | 77 | |
AK294945.1 | - | 8 - 9 | 1479 | 77 | |
BC070040.1 | - | 9 - 10 | 1948 | 77 | |
CR749673.1 | - | 8 - 9 | 1462 | 77 |