Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206846.1 | NP_001193775.1 | 10 - 11 | 3194 | 82 |
NM_018136.4 | NP_060606.3 | 10 - 11 | 3194 | 82 | |
GenBank mRNA | AF509326.1 | - | 10 - 11 | 2937 | 82 |
AK001380.1 | - | 6 - 7 | 869 | 82 | |
AK125107.1 | - | 10 - 11 | 3194 | 82 | |
AK226178.1 | - | 10 - 11 | 3127 | 82 | |
AY099891.1 | - | 8 - 9 | 1719 | 82 | |
AY099893.1 | - | 8 - 9 | 1719 | 82 | |
AY367065.1 | - | 10 - 11 | 2937 | 82 | |
AY971956.1 | - | 10 - 11 | 2937 | 82 | |
BC034607.1 | - | 6 - 7 | 813 | 82 |