Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42727529-42728030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDAN1
Entrez Gene ID
146059
Gene Name
codanin 1
Gene Aliases
CDA1, CDAI, CDAN1A, DLT, PRO1295
UniGene
Hs.599232
Gene Chromosome Location
Chr.15: 42723557-42737219 on Build GRCh38
Transcripts
NM_138477.2 XM_005254176.4 XM_011521270.2 XM_011521271.2 XM_011521274.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6046086 c.2876G>A p.S959N Chr.15: 42728026-42728026 on Build GRCh38
6387670 c.2956A>G p.R986G Chr.15: 42727761-42727761 on Build GRCh38
2187681 c.3045C>T p.R1015R Chr.15: 42727672-42727672 on Build GRCh38
6385268 c.3076C>A p.H1026N Chr.15: 42727641-42727641 on Build GRCh38
6034083 c.3025delG p.E1009fs*23 Chr.15: 42727692-42727692 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943832022 Chr.15: 42727645-42727645 on Build GRCh38
rs146070801 Chr.15: 42727722-42727722 on Build GRCh38
rs771314265 Chr.15: 42727655-42727655 on Build GRCh38
rs772513393 Chr.15: 42727701-42727701 on Build GRCh38
rs903818947 Chr.15: 42727920-42727920 on Build GRCh38
rs753780270 Chr.15: 42728002-42728002 on Build GRCh38
rs763819600 Chr.15: 42727999-42727999 on Build GRCh38
rs999212388 Chr.15: 42727579-42727579 on Build GRCh38
rs576998498 Chr.15: 42727576-42727576 on Build GRCh38
rs747974604 Chr.15: 42727692-42727692 on Build GRCh38
rs1043615882 Chr.15: 42727916-42727916 on Build GRCh38
rs781767396 Chr.15: 42727965-42727965 on Build GRCh38
rs763282803 Chr.15: 42727993-42727993 on Build GRCh38
rs1030736553 Chr.15: 42727580-42727580 on Build GRCh38
rs529199386 Chr.15: 42727806-42727806 on Build GRCh38
rs754935942 Chr.15: 42727691-42727691 on Build GRCh38
rs747575368 Chr.15: 42727803-42727803 on Build GRCh38
rs200419966 Chr.15: 42727799-42727799 on Build GRCh38
rs749885995 Chr.15: 42727710-42727711 on Build GRCh38
rs952191269 Chr.15: 42727835-42727835 on Build GRCh38
rs372998688 Chr.15: 42727946-42727946 on Build GRCh38
rs562819613 Chr.15: 42727629-42727629 on Build GRCh38
rs771026419 Chr.15: 42727994-42727994 on Build GRCh38
rs762526402 Chr.15: 42727647-42727647 on Build GRCh38
rs142276684 Chr.15: 42727698-42727698 on Build GRCh38
rs371757369 Chr.15: 42727688-42727688 on Build GRCh38
rs369919247 Chr.15: 42727733-42727733 on Build GRCh38
rs527704129 Chr.15: 42727926-42727926 on Build GRCh38
rs908047856 Chr.15: 42727867-42727867 on Build GRCh38
rs368795897 Chr.15: 42727985-42727985 on Build GRCh38
rs1019306872 Chr.15: 42728001-42728001 on Build GRCh38
rs1015790044 Chr.15: 42727699-42727699 on Build GRCh38
rs948078215 Chr.15: 42727899-42727899 on Build GRCh38
rs754121998 Chr.15: 42727676-42727676 on Build GRCh38
rs758732191 Chr.15: 42727672-42727672 on Build GRCh38
rs748594717 Chr.15: 42727970-42727970 on Build GRCh38
rs375186768 Chr.15: 42727797-42727797 on Build GRCh38
rs762364695 Chr.15: 42727593-42727610 on Build GRCh38
rs769454053 Chr.15: 42727570-42727570 on Build GRCh38
rs765548629 Chr.15: 42727728-42727728 on Build GRCh38
rs752802468 Chr.15: 42727614-42727614 on Build GRCh38
rs147554753 Chr.15: 42727673-42727673 on Build GRCh38
rs369437654 Chr.15: 42727608-42727608 on Build GRCh38
rs750125881 Chr.15: 42727942-42727942 on Build GRCh38
rs756972914 Chr.15: 42727769-42727769 on Build GRCh38
rs371248047 Chr.15: 42727552-42727553 on Build GRCh38
rs754219719 Chr.15: 42727582-42727599 on Build GRCh38
rs773569544 Chr.15: 42727821-42727821 on Build GRCh38
rs766672608 Chr.15: 42727584-42727584 on Build GRCh38
rs975420319 Chr.15: 42727642-42727642 on Build GRCh38
rs1044414655 Chr.15: 42727839-42727839 on Build GRCh38
rs1022098719 Chr.15: 42727569-42727569 on Build GRCh38
rs765671442 Chr.15: 42727600-42727601 on Build GRCh38
rs754811446 Chr.15: 42727731-42727731 on Build GRCh38
rs117946783 Chr.15: 42727782-42727782 on Build GRCh38
rs939420993 Chr.15: 42727875-42727875 on Build GRCh38
rs371073719 Chr.15: 42727898-42727898 on Build GRCh38
rs767514455 Chr.15: 42727610-42727610 on Build GRCh38
rs751454906 Chr.15: 42727729-42727729 on Build GRCh38
rs747687001 Chr.15: 42727640-42727640 on Build GRCh38
rs371854216 Chr.15: 42727663-42727663 on Build GRCh38
rs760375414 Chr.15: 42727998-42727998 on Build GRCh38
rs749882292 Chr.15: 42727640-42727641 on Build GRCh38
rs746228744 Chr.15: 42727700-42727700 on Build GRCh38
rs948707524 Chr.15: 42727695-42727695 on Build GRCh38
rs761702273 Chr.15: 42728007-42728007 on Build GRCh38
rs566135095 Chr.15: 42727717-42727717 on Build GRCh38
rs755653124 Chr.15: 42727696-42727696 on Build GRCh38
rs369116219 Chr.15: 42727671-42727671 on Build GRCh38
rs999348272 Chr.15: 42727780-42727780 on Build GRCh38
rs879634242 Chr.15: 42727743-42727743 on Build GRCh38
rs148259349 Chr.15: 42727991-42727991 on Build GRCh38
rs778052829 Chr.15: 42727623-42727623 on Build GRCh38
rs750301552 Chr.15: 42727624-42727624 on Build GRCh38
rs1012583217 Chr.15: 42727565-42727565 on Build GRCh38
rs375979132 Chr.15: 42727669-42727669 on Build GRCh38
rs759995430 Chr.15: 42727609-42727609 on Build GRCh38
rs758009560 Chr.15: 42727666-42727666 on Build GRCh38
rs746977550 Chr.15: 42727630-42727630 on Build GRCh38
rs761967705 Chr.15: 42727812-42727812 on Build GRCh38
rs752411544 Chr.15: 42727909-42727909 on Build GRCh38
rs767366816 Chr.15: 42727908-42727908 on Build GRCh38
rs548549298 Chr.15: 42727876-42727876 on Build GRCh38
rs199779909 Chr.15: 42727963-42727963 on Build GRCh38
rs764025922 Chr.15: 42727674-42727674 on Build GRCh38
rs199813215 Chr.15: 42727804-42727804 on Build GRCh38
rs200921566 Chr.15: 42727995-42727995 on Build GRCh38
rs541098619 Chr.15: 42727748-42727748 on Build GRCh38
rs748959489 Chr.15: 42727697-42727697 on Build GRCh38
rs776079135 Chr.15: 42727719-42727719 on Build GRCh38
rs765345774 Chr.15: 42727813-42727813 on Build GRCh38
rs975011454 Chr.15: 42727808-42727808 on Build GRCh38
rs753406643 Chr.15: 42727940-42727940 on Build GRCh38
rs948676667 Chr.15: 42727829-42727829 on Build GRCh38
rs752710811 Chr.15: 42727734-42727734 on Build GRCh38
rs201658845 Chr.15: 42727707-42727707 on Build GRCh38
rs539649120 Chr.15: 42727615-42727615 on Build GRCh38
rs757516734 Chr.15: 42727682-42727682 on Build GRCh38
rs764016082 Chr.15: 42727936-42727936 on Build GRCh38
rs780604551 Chr.15: 42727637-42727637 on Build GRCh38
rs764269200 Chr.15: 42727724-42727724 on Build GRCh38
rs779763549 Chr.15: 42727686-42727687 on Build GRCh38
rs572084618 Chr.15: 42727742-42727742 on Build GRCh38
rs747308319 Chr.15: 42727947-42727947 on Build GRCh38
rs748767255 Chr.15: 42727768-42727768 on Build GRCh38
rs763044755 Chr.15: 42727907-42727907 on Build GRCh38
rs564173096 Chr.15: 42727759-42727759 on Build GRCh38
rs764485826 Chr.15: 42727575-42727575 on Build GRCh38
rs762178142 Chr.15: 42727725-42727725 on Build GRCh38
rs751229677 Chr.15: 42727812-42727825 on Build GRCh38
rs752065246 Chr.15: 42727591-42727591 on Build GRCh38
rs145041909 Chr.15: 42727646-42727646 on Build GRCh38
rs745454394 Chr.15: 42727796-42727796 on Build GRCh38
rs375514582 Chr.15: 42727689-42727689 on Build GRCh38
rs983577111 Chr.15: 42727842-42727842 on Build GRCh38
rs995751855 Chr.15: 42727715-42727715 on Build GRCh38
rs920743807 Chr.15: 42727666-42727666 on Build GRCh38
rs1027156502 Chr.15: 42727912-42727912 on Build GRCh38

View Full Product Details