Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42727877-42728385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDAN1
Entrez Gene ID
146059
Gene Name
codanin 1
Gene Aliases
CDA1, CDAI, CDAN1A, DLT, PRO1295
UniGene
Hs.599232
Gene Chromosome Location
Chr.15: 42723557-42737219 on Build GRCh38
Transcripts
NM_138477.2 XM_005254176.4 XM_011521270.2 XM_011521271.2 XM_011521274.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6046086 c.2876G>A p.S959N Chr.15: 42728026-42728026 on Build GRCh38
4670586 c.2868+1G>A p.? Chr.15: 42728203-42728203 on Build GRCh38
5422806 c.2849C>T p.P950L Chr.15: 42728223-42728223 on Build GRCh38
6346472 c.2836C>A p.R946R Chr.15: 42728236-42728236 on Build GRCh38
470652 c.2921G>C p.C974S Chr.15: 42727981-42727981 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754625040 Chr.15: 42728159-42728159 on Build GRCh38
rs371165687 Chr.15: 42728295-42728295 on Build GRCh38
rs756751239 Chr.15: 42728183-42728183 on Build GRCh38
rs777603482 Chr.15: 42728159-42728161 on Build GRCh38
rs769707390 Chr.15: 42728057-42728058 on Build GRCh38
rs758691277 Chr.15: 42728276-42728276 on Build GRCh38
rs753780270 Chr.15: 42728002-42728002 on Build GRCh38
rs903818947 Chr.15: 42727920-42727920 on Build GRCh38
rs763819600 Chr.15: 42727999-42727999 on Build GRCh38
rs777946612 Chr.15: 42728035-42728035 on Build GRCh38
rs753071477 Chr.15: 42728357-42728357 on Build GRCh38
rs754334399 Chr.15: 42728243-42728243 on Build GRCh38
rs112759032 Chr.15: 42728092-42728092 on Build GRCh38
rs745898297 Chr.15: 42728167-42728186 on Build GRCh38
rs943973835 Chr.15: 42728346-42728346 on Build GRCh38
rs778130045 Chr.15: 42728186-42728186 on Build GRCh38
rs1043615882 Chr.15: 42727916-42727916 on Build GRCh38
rs763282803 Chr.15: 42727993-42727993 on Build GRCh38
rs781767396 Chr.15: 42727965-42727965 on Build GRCh38
rs770869536 Chr.15: 42728290-42728290 on Build GRCh38
rs931460177 Chr.15: 42728163-42728163 on Build GRCh38
rs200767055 Chr.15: 42728278-42728278 on Build GRCh38
rs1039455419 Chr.15: 42728196-42728196 on Build GRCh38
rs779555825 Chr.15: 42728015-42728015 on Build GRCh38
rs550681477 Chr.15: 42728123-42728123 on Build GRCh38
rs372998688 Chr.15: 42727946-42727946 on Build GRCh38
rs771026419 Chr.15: 42727994-42727994 on Build GRCh38
rs773023944 Chr.15: 42728211-42728211 on Build GRCh38
rs774069957 Chr.15: 42728294-42728294 on Build GRCh38
rs142833783 Chr.15: 42728235-42728235 on Build GRCh38
rs373134169 Chr.15: 42728059-42728059 on Build GRCh38
rs201757681 Chr.15: 42728232-42728232 on Build GRCh38
rs527704129 Chr.15: 42727926-42727926 on Build GRCh38
rs751163443 Chr.15: 42728019-42728019 on Build GRCh38
rs1019306872 Chr.15: 42728001-42728001 on Build GRCh38
rs368795897 Chr.15: 42727985-42727985 on Build GRCh38
rs377567798 Chr.15: 42728335-42728335 on Build GRCh38
rs62020665 Chr.15: 42728126-42728126 on Build GRCh38
rs764432820 Chr.15: 42728030-42728030 on Build GRCh38
rs749746339 Chr.15: 42728200-42728200 on Build GRCh38
rs140275867 Chr.15: 42728033-42728033 on Build GRCh38
rs748594717 Chr.15: 42727970-42727970 on Build GRCh38
rs754747407 Chr.15: 42728043-42728044 on Build GRCh38
rs559228588 Chr.15: 42728264-42728264 on Build GRCh38
rs1048559811 Chr.15: 42728197-42728197 on Build GRCh38
rs114779238 Chr.15: 42728236-42728236 on Build GRCh38
rs747910800 Chr.15: 42728051-42728052 on Build GRCh38
rs117597169 Chr.15: 42728100-42728100 on Build GRCh38
rs749321986 Chr.15: 42728037-42728037 on Build GRCh38
rs748197591 Chr.15: 42728054-42728054 on Build GRCh38
rs750125881 Chr.15: 42727942-42727942 on Build GRCh38
rs916471417 Chr.15: 42728120-42728120 on Build GRCh38
rs1026619957 Chr.15: 42728209-42728209 on Build GRCh38
rs769503711 Chr.15: 42728284-42728284 on Build GRCh38
rs749229757 Chr.15: 42728289-42728289 on Build GRCh38
rs772262599 Chr.15: 42728205-42728205 on Build GRCh38
rs747164189 Chr.15: 42728282-42728282 on Build GRCh38
rs776454812 Chr.15: 42728207-42728207 on Build GRCh38
rs759103652 Chr.15: 42728310-42728310 on Build GRCh38
rs938755549 Chr.15: 42728131-42728131 on Build GRCh38
rs757734894 Chr.15: 42728245-42728245 on Build GRCh38
rs187355078 Chr.15: 42728340-42728340 on Build GRCh38
rs952238284 Chr.15: 42728067-42728067 on Build GRCh38
rs1052326712 Chr.15: 42728148-42728148 on Build GRCh38
rs774451396 Chr.15: 42728041-42728041 on Build GRCh38
rs1037087427 Chr.15: 42728143-42728143 on Build GRCh38
rs758112958 Chr.15: 42728014-42728014 on Build GRCh38
rs765058557 Chr.15: 42728065-42728065 on Build GRCh38
rs781030207 Chr.15: 42728049-42728050 on Build GRCh38
rs374090305 Chr.15: 42728299-42728299 on Build GRCh38
rs925269677 Chr.15: 42728316-42728316 on Build GRCh38
rs756449578 Chr.15: 42728239-42728239 on Build GRCh38
rs556046335 Chr.15: 42728099-42728099 on Build GRCh38
rs760375414 Chr.15: 42727998-42727998 on Build GRCh38
rs761702273 Chr.15: 42728007-42728007 on Build GRCh38
rs1041106624 Chr.15: 42728219-42728219 on Build GRCh38
rs148259349 Chr.15: 42727991-42727991 on Build GRCh38
rs768041338 Chr.15: 42728171-42728171 on Build GRCh38
rs556528780 Chr.15: 42728050-42728050 on Build GRCh38
rs150438471 Chr.15: 42728204-42728204 on Build GRCh38
rs145148854 Chr.15: 42728203-42728203 on Build GRCh38
rs765844268 Chr.15: 42728070-42728070 on Build GRCh38
rs1048527088 Chr.15: 42728341-42728341 on Build GRCh38
rs753079849 Chr.15: 42728182-42728182 on Build GRCh38
rs146535176 Chr.15: 42728115-42728115 on Build GRCh38
rs973165559 Chr.15: 42728317-42728317 on Build GRCh38
rs781653078 Chr.15: 42728026-42728026 on Build GRCh38
rs969290235 Chr.15: 42728291-42728291 on Build GRCh38
rs772849591 Chr.15: 42728068-42728068 on Build GRCh38
rs570121114 Chr.15: 42728343-42728343 on Build GRCh38
rs565486675 Chr.15: 42728097-42728097 on Build GRCh38
rs377606247 Chr.15: 42728149-42728149 on Build GRCh38
rs770017185 Chr.15: 42728090-42728090 on Build GRCh38
rs751198894 Chr.15: 42728158-42728158 on Build GRCh38
rs541034913 Chr.15: 42728112-42728112 on Build GRCh38
rs138228278 Chr.15: 42728231-42728231 on Build GRCh38
rs752411544 Chr.15: 42727909-42727909 on Build GRCh38
rs747935657 Chr.15: 42728210-42728210 on Build GRCh38
rs767366816 Chr.15: 42727908-42727908 on Build GRCh38
rs199779909 Chr.15: 42727963-42727963 on Build GRCh38
rs546806428 Chr.15: 42728147-42728147 on Build GRCh38
rs139032045 Chr.15: 42728270-42728270 on Build GRCh38
rs201343155 Chr.15: 42728048-42728048 on Build GRCh38
rs200921566 Chr.15: 42727995-42727995 on Build GRCh38
rs759300971 Chr.15: 42728233-42728233 on Build GRCh38
rs140543685 Chr.15: 42728334-42728334 on Build GRCh38
rs141236475 Chr.15: 42728130-42728130 on Build GRCh38
rs753406643 Chr.15: 42727940-42727940 on Build GRCh38
rs375929824 Chr.15: 42728285-42728285 on Build GRCh38
rs547440410 Chr.15: 42728020-42728020 on Build GRCh38
rs570313166 Chr.15: 42728063-42728063 on Build GRCh38
rs764016082 Chr.15: 42727936-42727936 on Build GRCh38
rs747308319 Chr.15: 42727947-42727947 on Build GRCh38
rs763044755 Chr.15: 42727907-42727907 on Build GRCh38
rs943904295 Chr.15: 42728167-42728167 on Build GRCh38
rs370304543 Chr.15: 42728038-42728038 on Build GRCh38
rs772115656 Chr.15: 42728300-42728300 on Build GRCh38
rs762379774 Chr.15: 42728069-42728069 on Build GRCh38
rs749881716 Chr.15: 42728010-42728010 on Build GRCh38
rs762904904 Chr.15: 42728223-42728223 on Build GRCh38
rs1027156502 Chr.15: 42727912-42727912 on Build GRCh38

View Full Product Details