Assay Design



Gene Transcripts

Gene Symbol
SASH1
Entrez Gene ID
23328
Gene Name
SAM and SH3 domain containing 1
Gene Aliases
SH3D6A, dJ323M4, dJ323M4.1
Chromosome Location
Chr.6: 148193440 - 148552049 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015278.3 NP_056093.3 10 - 11 1686 113
XM_005266875.1 XP_005266932.1 3 - 4 912 113
XM_011535640.1 XP_011533942.1 10 - 11 1129 113
XM_017010607.1 XP_016866096.1 11 - 12 1309 113
GenBank mRNA AB018333.1 - 10 - 11 1429 113
AJ507735.1 - 10 - 11 1217 113
AK024403.1 - 4 - 5 671 113
AK025495.1 - 10 - 11 1686 113
AK300403.1 - 3 - 4 596 113
BC063279.1 - 10 - 11 1155 113

View Full Product Details